The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008718.2 | NP_032744.1 | 11 - 12 | 1505 | 71 |
GenBank mRNA | AK158830.1 | - | 9 - 10 | 2162 | 71 |
BC132113.1 | - | 10 - 11 | 1340 | 71 | |
BC137863.1 | - | 10 - 11 | 1340 | 71 | |
U77967.1 | - | 10 - 11 | 1373 | 71 |