Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cryba1
Entrez Gene ID
12957
Gene Name
crystallin, beta A1
Gene Aliases
BA3/A1, Cryb
Chromosome Location
Chr.11: 77718732 - 77725781 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009965.2 NP_034095.1 5 - 6 507 120
GenBank mRNA AJ239052.1 - 5 - 6 507 120
AK084145.1 - 2 - 3 1084 120
BC107342.2 - 5 - 6 507 120
J00378.1 - 2 - 3 280 120
V00728.1 - 3 - 4 399 120

View Full Product Details