Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zfyve21
Entrez Gene ID
68520
Gene Name
zinc finger, FYVE domain containing 21
Gene Aliases
1110013H04Rik, AW558780, C85416
Chromosome Location
Chr.12: 111814170 - 111828388 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026752.4 NP_081028.3 4 - 5 500 85
GenBank mRNA AK003661.1 - 4 - 5 397 85
AK139356.1 - - 435 85
AK155300.1 - 4 - 5 501 85
AK171099.1 - 4 - 5 441 85
AK192012.1 - 2 - 3 110 85
AK197357.1 - 2 - 3 111 85
BC019521.1 - 4 - 5 430 85
BC132248.1 - 4 - 5 400 85
BC132250.1 - 4 - 5 400 85

View Full Product Details