The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164607.1 | NP_001158079.1 | 7 - 8 | 1082 | 109 |
NM_001164608.1 | NP_001158080.1 | 6 - 7 | 989 | 109 | |
NM_026859.3 | NP_081135.3 | 6 - 7 | 1036 | 109 | |
GenBank mRNA | AK004406.1 | - | 6 - 7 | 709 | 109 |
AK042913.1 | - | 6 - 7 | 930 | 109 | |
AK154047.1 | - | 6 - 7 | 979 | 109 | |
BC016260.1 | - | 6 - 7 | 1024 | 109 |