The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029770.2 | NP_084046.2 | 15 - 16 | 2905 | 67 |
GenBank mRNA | AJ487853.1 | - | 15 - 16 | 2808 | 67 |
AK018177.1 | - | 15 - 16 | 2905 | 67 | |
AK147603.1 | - | 14 - 15 | 2767 | 67 | |
AK154271.1 | - | 15 - 16 | 2905 | 67 | |
AK154287.1 | - | 15 - 16 | 2909 | 67 | |
AK158284.1 | - | 15 - 16 | 2905 | 67 | |
BC048162.1 | - | 14 - 15 | 2679 | 67 | |
BC057560.1 | - | 14 - 15 | 2679 | 67 |