Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fbxl14
Entrez Gene ID
101358
Gene Name
F-box and leucine-rich repeat protein 14
Gene Aliases
AW322056, Fbx14l
Chromosome Location
Chr.6: 119479668 - 119483886 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133940.3 NP_598701.2 1 - 1 1761 102
GenBank mRNA AK036124.1 - 1 - 1 1658 102
AK084506.1 - 1 - 1 1648 102
AK155154.1 - 1 - 1 1644 102
BC006913.1 - 1 - 1 1350 102
BC021329.1 - 1 - 1 1654 102

View Full Product Details