Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Yipf2
Entrez Gene ID
74766
Gene Name
Yip1 domain family, member 2
Gene Aliases
1300010K09Rik
Chromosome Location
Chr.9: 21588682 - 21592831 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205157.1 NP_001192086.1 6 - 7 611 74
NM_001205158.1 NP_001192087.1 6 - 7 601 74
NM_138303.2 NP_612176.1 5 - 6 749 74
NR_038062.1 5 - 6 550 74
GenBank mRNA AK004959.1 - 5 - 6 535 74
AK033705.1 - 6 - 7 560 74
AK133631.1 - 6 - 7 571 74
BC002282.1 - 5 - 6 664 74
BC003289.1 - 6 - 7 542 74

View Full Product Details