The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_139134.3 | NP_624360.2 | 5 - 6 | 1152 | 76 |
GenBank mRNA | AF311699.1 | - | 5 - 6 | 975 | 76 |
AK014255.1 | - | 5 - 6 | 863 | 76 | |
AK140910.1 | - | 5 - 6 | 1153 | 76 | |
BC117071.1 | - | 5 - 6 | 823 | 76 | |
BC117073.1 | - | 5 - 6 | 823 | 76 | |
BC144738.1 | - | 5 - 6 | 808 | 76 |