The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026619.2 | NP_080895.2 | 2 - 3 | 216 | 103 |
NM_030051.1 | NP_084327.1 | 2 - 3 | 169 | 103 | |
GenBank mRNA | AK018953.1 | - | 2 - 3 | 207 | 103 |
AK019582.1 | - | 2 - 3 | 135 | 103 | |
AK075753.1 | - | 2 - 3 | 217 | 103 | |
AK077086.1 | - | 2 - 3 | 170 | 103 |