The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027494.3 | NP_081770.3 | 10 - 11 | 1141 | 107 |
GenBank mRNA | AK017857.1 | - | 10 - 11 | 1037 | 107 |
AK032349.1 | - | 10 - 11 | 1118 | 107 | |
AK033469.1 | - | 10 - 11 | 1091 | 107 | |
AK156770.1 | - | 10 - 11 | 1113 | 107 | |
AK166990.1 | - | 10 - 11 | 1048 | 107 | |
AK167126.1 | - | 10 - 11 | 1046 | 107 | |
AK167736.1 | - | 10 - 11 | 1048 | 107 | |
BC013555.1 | - | 9 - 10 | 793 | 107 |