The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028131.3 | NP_082407.1 | 7 - 8 | 691 | 63 |
GenBank mRNA | AK012097.1 | - | 7 - 8 | 692 | 63 |
AK219823.1 | - | 1 - 2 | 75 | 63 | |
BC027119.1 | - | 5 - 6 | 506 | 63 | |
BC058243.1 | - | 5 - 6 | 602 | 63 |