The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008409.2 | NP_032435.2 | 5 - 6 | 848 | 114 |
GenBank mRNA | AK014024.1 | - | 5 - 6 | 616 | 114 |
AK051956.1 | - | 5 - 6 | 827 | 114 | |
AK135229.1 | - | 5 - 6 | 800 | 114 | |
AK167351.1 | - | 5 - 6 | 849 | 114 | |
BC031420.1 | - | 5 - 6 | 815 | 114 | |
L38971.1 | - | 5 - 6 | 851 | 114 |