The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267621.1 | NP_001254550.1 | 5 - 6 | 1218 | 70 |
NM_010278.2 | NP_034408.1 | 4 - 5 | 1430 | 70 | |
GenBank mRNA | BC130219.1 | - | 3 - 4 | 615 | 70 |
BC152328.1 | - | 3 - 4 | 506 | 70 | |
BU698904.1 | - | 2 - 3 | 246 | 70 | |
U58972.1 | - | 4 - 5 | 1405 | 70 | |
U78312.1 | - | 4 - 5 | 1430 | 70 |