The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013478.2 | NP_038506.2 | 3 - 4 | 641 | 70 |
GenBank mRNA | AK005051.1 | - | 3 - 4 | 627 | 70 |
AY248694.1 | - | 3 - 4 | 602 | 70 | |
BC061646.1 | - | 3 - 4 | 623 | 70 | |
D21059.1 | - | 3 - 4 | 602 | 70 |