Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Frg1
Entrez Gene ID
14300
Gene Name
FSHD region gene 1
Gene Aliases
-
Chromosome Location
Chr.8: 41397454 - 41417118 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013522.3 NP_038550.2 1 - 2 214 85
GenBank mRNA AK077107.1 - 1 - 2 141 85
AK079229.1 - 1 - 2 106 85
BC002027.1 - 1 - 2 170 85
BE655768.1 - 1 - 2 214 85
CB573470.1 - 1 - 2 176 85
U62105.1 - 1 - 2 173 85

View Full Product Details