The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204165.1 | NP_001191094.1 | 20 - 21 | 2003 | 69 |
NM_001204166.1 | NP_001191095.1 | 17 - 18 | 1895 | 69 | |
NM_019688.2 | NP_062662.1 | 19 - 20 | 1949 | 69 | |
GenBank mRNA | AB021132.1 | - | 20 - 21 | 1978 | 69 |
AB037668.1 | - | 11 - 12 | 1338 | 69 | |
AB445011.1 | - | 17 - 18 | 1887 | 69 | |
AF115480.1 | - | 19 - 20 | 1877 | 69 | |
AK004874.1 | - | 11 - 12 | 995 | 69 | |
AK220281.1 | - | 20 - 21 | 2003 | 69 | |
BC139822.1 | - | 20 - 21 | 1935 | 69 |