The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285463.1 | NP_001272392.1 | 8 - 9 | 823 | 61 |
NM_001285473.1 | NP_001272402.1 | 4 - 5 | 413 | 61 | |
NM_139150.5 | NP_631889.1 | 6 - 7 | 696 | 61 | |
NR_104344.1 | 4 - 5 | 413 | 61 | ||
GenBank mRNA | AB221600.1 | - | 5 - 6 | 479 | 61 |
AF454947.1 | - | 6 - 7 | 665 | 61 | |
BC138644.1 | - | 5 - 6 | 602 | 61 | |
BC138645.1 | - | 5 - 6 | 602 | 61 | |
BC145350.1 | - | 5 - 6 | 598 | 61 | |
EU170502.1 | - | 4 - 5 | 394 | 61 | |
EU170503.1 | - | 4 - 5 | 394 | 61 |