Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mgat4b
Entrez Gene ID
103534
Gene Name
mannoside acetylglucosaminyltransferase 4, isoenzyme B
Gene Aliases
AA407995, GnTIVb
Chromosome Location
Chr.11: 50225335 - 50235103 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145926.3 NP_666038.3 11 - 12 1651 72
GenBank mRNA AB053218.1 - 11 - 12 1353 72
AK155336.1 - 11 - 12 1652 72
BC026638.1 - 6 - 7 759 72
BC031613.1 - 5 - 6 629 72
CX206051.1 - 3 - 4 410 72

View Full Product Details