Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Depdc7
Entrez Gene ID
211896
Gene Name
DEP domain containing 7
Gene Aliases
AV216087
Chromosome Location
Chr.2: 104721784 - 104742801 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144804.1 NP_659053.1 3 - 4 632 77
GenBank mRNA AK134830.1 - 3 - 4 652 77
AK144066.1 - 3 - 4 645 77
AK162218.1 - 3 - 4 655 77
AK165504.1 - 3 - 4 645 77
BC013499.1 - 3 - 4 632 77

View Full Product Details