Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clptm1l
Entrez Gene ID
218335
Gene Name
CLPTM1-like
Gene Aliases
C130052I12Rik, Crr9
Chromosome Location
Chr.13: 73604002 - 73620639 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146047.2 NP_666159.2 4 - 5 841 68
GenBank mRNA AK032295.1 - 4 - 5 804 68
AK048357.1 - 4 - 5 738 68
AK165939.1 - 4 - 5 701 68
AK192551.1 - 2 - 3 357 68
BC026562.1 - 4 - 5 542 68

View Full Product Details