The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164357.1 | NP_001157829.1 | 6 - 7 | 1046 | 92 |
NM_001164358.1 | NP_001157830.1 | 7 - 8 | 1301 | 92 | |
NM_001290558.1 | NP_001277487.1 | 6 - 7 | 915 | 92 | |
NM_146118.3 | NP_666230.2 | 7 - 8 | 1082 | 92 | |
GenBank mRNA | AK049392.1 | - | 6 - 7 | 1022 | 92 |
AK082756.1 | - | 6 - 7 | 1003 | 92 | |
AK122568.2 | - | 7 - 8 | 1266 | 92 | |
AK132204.1 | - | 6 - 7 | 1000 | 92 | |
BC019978.1 | - | 6 - 7 | 854 | 92 | |
BC022114.1 | - | 6 - 7 | 852 | 92 | |
BC037109.1 | - | 7 - 8 | 1042 | 92 | |
BC066998.1 | - | 7 - 8 | 1269 | 92 |