Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arx
Entrez Gene ID
11878
Gene Name
aristaless related homeobox
Gene Aliases
-
Chromosome Location
Chr.X: 93286450 - 93298357 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB006103.2 - 5 - 6 1545 104
BC052033.1 - 4 - 5 1539 104

View Full Product Details