The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252568.2 | NP_001239497.1 | 2 - 3 | 507 | 71 |
NM_001252570.1 | NP_001239499.1 | 1 - 2 | 128 | 71 | |
NM_001252571.1 | NP_001239500.1 | 1 - 2 | 128 | 71 | |
NM_001281829.1 | NP_001268758.1 | 2 - 3 | 154 | 71 | |
NM_172267.4 | NP_758471.1 | 2 - 3 | 507 | 71 | |
GenBank mRNA | AI182929.1 | - | 2 - 3 | 129 | 71 |
AK005293.1 | - | 2 - 3 | 508 | 71 | |
AK046118.1 | - | 1 - 2 | 97 | 71 | |
AK138607.1 | - | 2 - 3 | 182 | 71 | |
BC039982.1 | - | 2 - 3 | 149 | 71 | |
BC132271.1 | - | 2 - 3 | 118 | 71 | |
BC132273.1 | - | 2 - 3 | 118 | 71 | |
BC145356.1 | - | 2 - 3 | 118 | 71 | |
BC171925.1 | - | 2 - 3 | 118 | 71 | |
BQ829083.1 | - | 1 - 2 | 124 | 71 | |
BY196857.1 | - | 2 - 3 | 156 | 71 | |
BY344206.1 | - | 2 - 3 | 161 | 71 |