Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cpt1a
Entrez Gene ID
12894
Gene Name
carnitine palmitoyltransferase 1a, liver
Gene Aliases
C730027G07, CPTI, Cpt1
Chromosome Location
Chr.19: 3322326 - 3385735 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013495.2 NP_038523.2 15 - 16 1966 84
GenBank mRNA AF017175.1 - - 84
AK077454.1 - 15 - 16 1920 84
AK128925.1 - 15 - 16 1920 84
AK133472.1 - 15 - 16 1921 84
AK136487.1 - 15 - 16 1920 84
AK147770.1 - 15 - 16 1921 84
AK149688.1 - 15 - 16 1917 84
BC038395.1 - 14 - 15 1901 84
BC046383.1 - 14 - 15 1901 84
BC054791.1 - 15 - 16 1947 84
D49472.1 - 14 - 15 1893 84

View Full Product Details