Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmco3
Entrez Gene ID
234076
Gene Name
transmembrane and coiled-coil domains 3
Gene Aliases
B230339H12Rik, C87304
Chromosome Location
Chr.8: 13288013 - 13322924 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172282.2 NP_758486.1 7 - 8 1678 63
GenBank mRNA AK033499.1 - 7 - 8 1439 63
AK088866.1 - 7 - 8 1462 63
AK148252.1 - 7 - 8 1481 63
BC039955.1 - 7 - 8 1424 63

View Full Product Details