The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172499.2 | NP_766087.1 | 5 - 6 | 643 | 77 |
GenBank mRNA | AK029874.1 | - | 5 - 6 | 644 | 77 |
AK169911.1 | - | 6 - 7 | 804 | 77 | |
BC057633.1 | - | 5 - 6 | 649 | 77 |