The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172516.4 | NP_766104.2 | 6 - 7 | 1964 | 66 |
GenBank mRNA | AK043199.1 | - | 1 - 2 | 833 | 66 |
AK050542.1 | - | 2 - 3 | 210 | 66 | |
AK083696.1 | - | 6 - 7 | 1862 | 66 | |
AK132119.1 | - | 6 - 7 | 1839 | 66 | |
AK145449.1 | - | 6 - 7 | 1809 | 66 | |
AK172954.1 | - | 6 - 7 | 1810 | 66 | |
AY208852.1 | - | 6 - 7 | 1811 | 66 | |
AY429676.1 | - | 6 - 7 | 1850 | 66 | |
BC019133.1 | - | 4 - 5 | 599 | 66 | |
BC089380.1 | - | 3 - 4 | 381 | 66 |