The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024458.3 | NP_001019629.2 | 1 - 2 | 203 | 64 |
NM_001102444.1 | NP_001095914.1 | 1 - 2 | 203 | 64 | |
NM_013457.3 | NP_038485.1 | 1 - 2 | 203 | 64 | |
GenBank mRNA | AF096839.1 | - | 1 - 2 | 101 | 64 |
AK054558.1 | - | 1 - 2 | 55 | 64 | |
AK079960.1 | - | 1 - 2 | 58 | 64 | |
CD349465.1 | - | 1 - 2 | 203 | 64 |