Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Stk36
Entrez Gene ID
269209
Gene Name
serine/threonine kinase 36
Gene Aliases
1700112N14Rik, B930045J24, FU, Fused, mKIAA1278
Chromosome Location
Chr.1: 74601450 - 74636894 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175031.3 NP_778196.2 1 - 2 164 106
GenBank mRNA AK007188.1 - 1 - 2 107 106
AK029842.1 - 1 - 2 140 106
AK173142.1 - 1 - 2 124 106
BC043103.1 - 1 - 2 116 106
BC049721.1 - 1 - 2 90 106
CK633987.1 - 1 - 2 168 105

View Full Product Details