Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Reps2
Entrez Gene ID
194590
Gene Name
RALBP1 associated Eps domain containing protein 2
Gene Aliases
POB1
Chromosome Location
Chr.X: 162411952 - 162643705 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290633.1 NP_001277562.1 8 - 9 1222 68
NM_178256.4 NP_839987.2 8 - 9 1225 68
GenBank mRNA AK147418.1 - 8 - 9 873 68
AK147432.1 - 5 - 6 504 68
AY263368.1 - 8 - 9 1170 68
BC139012.1 - 8 - 9 1101 68
BC145500.1 - 8 - 9 1098 68

View Full Product Details