The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178395.3 | NP_848482.1 | 11 - 12 | 1284 | 86 |
GenBank mRNA | AK018284.1 | - | - | 1255 | 86 |
AK030662.1 | - | 11 - 12 | 1285 | 86 | |
AK046533.1 | - | 11 - 12 | 1284 | 86 | |
AK136296.1 | - | 11 - 12 | 1282 | 86 | |
BC117761.1 | - | 12 - 13 | 1046 | 86 | |
BC127156.1 | - | 12 - 13 | 1118 | 86 |