The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001162485.1 | NP_001155957.1 | 3 - 4 | 419 | 68 |
NM_178408.3 | NP_848495.2 | 3 - 4 | 419 | 68 | |
NR_027876.1 | 3 - 4 | 419 | 68 | ||
GenBank mRNA | AK088008.1 | - | 3 - 4 | 284 | 68 |
AK170018.1 | - | 3 - 4 | 293 | 68 | |
BC004091.1 | - | 3 - 4 | 314 | 68 | |
BC018501.1 | - | 3 - 4 | 301 | 68 |