The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001162485.1 | NP_001155957.1 | 7 - 8 | 1369 | 69 |
NM_178408.3 | NP_848495.2 | 7 - 8 | 1366 | 69 | |
NR_027876.1 | 6 - 7 | 1408 | 69 | ||
GenBank mRNA | AK088008.1 | - | 6 - 7 | 1273 | 69 |
AK170018.1 | - | 7 - 8 | 1243 | 69 | |
BC004091.1 | - | 7 - 8 | 1264 | 69 | |
BC018501.1 | - | 7 - 8 | 1248 | 69 |