The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178624.6 | NP_848739.1 | 3 - 4 | 177 | 79 |
GenBank mRNA | AK013142.1 | - | 4 - 5 | 311 | 79 |
AK039010.1 | - | 3 - 4 | 177 | 79 | |
AK044693.1 | - | 3 - 4 | 174 | 79 | |
AK045742.1 | - | 3 - 4 | 164 | 79 | |
AK089994.1 | - | 3 - 4 | 153 | 79 | |
AK141177.1 | - | 3 - 4 | 167 | 79 | |
AK163472.1 | - | 3 - 4 | 173 | 79 | |
BC096582.1 | - | 3 - 4 | 163 | 79 | |
BC145665.1 | - | 3 - 4 | 157 | 79 | |
BC145998.1 | - | 3 - 4 | 157 | 79 |