The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178683.4 | NP_848798.1 | 10 - 11 | 1518 | 148 |
GenBank mRNA | AK036550.1 | - | 10 - 11 | 1477 | 148 |
AK043283.1 | - | - | 1475 | 148 | |
AK077676.1 | - | 10 - 11 | 1519 | 148 | |
AK083480.1 | - | 10 - 11 | 1514 | 148 | |
BC048246.1 | - | 3 - 4 | 550 | 148 | |
BC152330.1 | - | 10 - 11 | 1523 | 148 | |
BC152331.1 | - | 10 - 11 | 1523 | 148 |