The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177566.3 | NP_808234.2 | 7 - 8 | 1662 | 103 |
GenBank mRNA | AK051749.1 | - | 4 - 5 | 792 | 103 |
AK173061.1 | - | 3 - 4 | 367 | 103 | |
BC089365.1 | - | 7 - 8 | 1662 | 103 | |
BC116343.1 | - | 7 - 8 | 1638 | 103 | |
BC116344.1 | - | 7 - 8 | 1638 | 103 |