The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177680.3 | NP_808348.2 | 9 - 10 | 1428 | 89 |
GenBank mRNA | AB221619.1 | - | 1 - 2 | 52 | 89 |
AI121974.1 | - | 2 - 3 | 163 | 89 | |
AK173312.1 | - | 6 - 7 | 691 | 89 | |
BC022697.1 | - | 2 - 3 | 221 | 89 | |
BC037593.1 | - | 3 - 4 | 321 | 89 | |
BC044650.1 | - | 7 - 8 | 1246 | 89 | |
BC048817.1 | - | 2 - 3 | 206 | 89 | |
CO424360.1 | - | 4 - 5 | 373 | 89 |