Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rft1
Entrez Gene ID
328370
Gene Name
RFT1 homolog (S. cerevisiae)
Gene Aliases
D930025H04
Chromosome Location
Chr.14: 30654341 - 30691317 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177815.3 NP_808483.2 4 - 5 492 73
GenBank mRNA AK086390.1 - 4 - 5 481 73
BC082539.1 - 4 - 5 460 73
BC116368.2 - 4 - 5 473 73
BC116369.2 - 4 - 5 473 73
BC141275.1 - 4 - 5 486 73
BC145606.1 - 4 - 5 486 73

View Full Product Details