Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dppa1
Entrez Gene ID
347708
Gene Name
developmental pluripotency associated 1
Gene Aliases
C77727
Chromosome Location
Chr.11: 46607010 - 46632233 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163358.1 NP_001156830.1 3 - 4 165 68
NM_178247.3 NP_839978.1 4 - 5 251 68
GenBank mRNA AF490345.1 - 4 - 5 237 68
AK145642.1 - 4 - 5 252 68
AK166752.1 - 4 - 5 249 68
BC115806.1 - 3 - 4 192 68
BC115901.1 - 3 - 4 189 68

View Full Product Details