The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277229.1 | NP_001264158.1 | 1 - 2 | 176 | 78 |
NM_001277230.1 | NP_001264159.1 | 1 - 2 | 176 | 78 | |
NM_153398.3 | NP_700447.2 | 1 - 2 | 176 | 78 | |
NR_102360.1 | 1 - 2 | 176 | 78 | ||
GenBank mRNA | AF263010.2 | - | 1 - 2 | 99 | 78 |
AK054097.1 | - | 1 - 2 | 132 | 78 | |
AK146986.1 | - | 1 - 2 | 152 | 78 | |
AK154467.1 | - | 1 - 2 | 152 | 78 | |
AK169260.1 | - | 1 - 2 | 123 | 78 | |
AY210402.1 | - | 1 - 2 | 150 | 78 | |
BC050933.1 | - | 1 - 2 | 138 | 78 | |
BC055367.1 | - | 1 - 2 | 119 | 78 |