The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027179.1 | NP_081455.1 | 9 - 10 | 759 | 143 |
NM_134118.5 | NP_598879.1 | 10 - 11 | 804 | 143 | |
GenBank mRNA | AK002384.1 | - | 10 - 11 | 774 | 143 |
AK010513.1 | - | 9 - 10 | 695 | 143 | |
AK010984.1 | - | 10 - 11 | 778 | 143 | |
AK013558.1 | - | 10 - 11 | 756 | 142 | |
AK020708.1 | - | 9 - 10 | 755 | 143 | |
AK171718.1 | - | 5 - 6 | 511 | 143 | |
AK172506.1 | - | 5 - 6 | 489 | 143 | |
BC019984.1 | - | 10 - 11 | 790 | 143 | |
BC094046.1 | - | 10 - 11 | 904 | 143 |