The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113179.1 | NP_001106650.1 | 24 - 25 | 3101 | 92 |
NM_009772.2 | NP_033902.2 | 24 - 25 | 3098 | 92 | |
GenBank mRNA | AF002823.1 | - | 24 - 25 | 3044 | 92 |
AK042932.1 | - | 24 - 25 | 3100 | 92 | |
AK144213.1 | - | 24 - 25 | 3110 | 92 | |
BC029116.1 | - | 24 - 25 | 3067 | 92 | |
U89795.1 | - | 24 - 25 | 3116 | 92 |