Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Epn3
Entrez Gene ID
71889
Gene Name
epsin 3
Gene Aliases
2310022G12Rik
Chromosome Location
Chr.11: 94489599 - 94499974 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027984.3 NP_082260.1 7 - 8 1650 115
GenBank mRNA AK009469.1 - 6 - 7 1030 115
AK158031.1 - 6 - 7 1528 115
BC016454.1 - 7 - 8 1599 115

View Full Product Details