Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lrrn4
Entrez Gene ID
320974
Gene Name
leucine rich repeat neuronal 4
Gene Aliases
B430119L13Rik, Nlrr4
Chromosome Location
Chr.2: 132866194 - 132894009 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177303.4 NP_796277.2 1 - 2 70 62
GenBank mRNA AK080889.1 - 1 - 2 71 62
AK085246.1 - 1 - 2 100 62
AY325108.1 - 1 - 2 53 62

View Full Product Details