The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008219.3 | NP_032245.1 | 2 - 3 | 365 | 92 |
GenBank mRNA | AK011296.1 | - | 2 - 3 | 366 | 92 |
AK011309.1 | - | 2 - 3 | 364 | 92 | |
AK012442.1 | - | 2 - 3 | 366 | 92 | |
AK012684.1 | - | 2 - 3 | 366 | 92 | |
AK028208.1 | - | 2 - 3 | 368 | 92 | |
AK160642.1 | - | 2 - 3 | 365 | 92 | |
BC052008.1 | - | 2 - 3 | 340 | 92 | |
BC089456.1 | - | 2 - 3 | 340 | 92 | |
M26894.1 | - | 2 - 3 | 292 | 92 |