The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027321.3 | NP_081597.1 | 4 - 5 | 609 | 149 |
NM_175413.3 | NP_780622.1 | 4 - 5 | 609 | 149 | |
GenBank mRNA | AK008119.1 | - | 4 - 5 | 577 | 149 |
AK035072.1 | - | 4 - 5 | 611 | 149 | |
AK039277.1 | - | 4 - 5 | 606 | 149 | |
AK046671.1 | - | 4 - 5 | 505 | 149 | |
AK052159.1 | - | 4 - 5 | 561 | 149 | |
AK079125.1 | - | 4 - 5 | 587 | 149 | |
BC119529.2 | - | 4 - 5 | 558 | 149 | |
BC119530.1 | - | 4 - 5 | 559 | 149 |