Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phf6
Entrez Gene ID
70998
Gene Name
PHD finger protein 6
Gene Aliases
2700007B13Rik, 4931428F02Rik, mKIAA1823
Chromosome Location
Chr.X: 52912214 - 52956961 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290546.1 NP_001277475.1 6 - 7 817 118
NM_027642.2 NP_081918.1 8 - 9 1103 118
GenBank mRNA AK016480.1 - 8 - 9 1104 118
AK028498.1 - 6 - 7 739 118
AK122560.1 - 8 - 9 1033 118
AK141804.1 - 8 - 9 1046 118
AK163345.1 - 8 - 9 1057 118
BC043127.1 - 8 - 9 1051 118
BC055330.1 - 8 - 9 1046 118
BC057374.1 - 8 - 9 1009 118

View Full Product Details