The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183136.2 | NP_898959.2 | 1 - 2 | 203 | 66 |
GenBank mRNA | AK021276.1 | - | 1 - 2 | 186 | 66 |
BC048588.1 | - | 1 - 2 | 203 | 66 | |
CN843460.1 | - | 1 - 2 | 149 | 66 | |
DQ437329.1 | - | 1 - 2 | 185 | 66 |