The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183165.3 | NP_898988.2 | 11 - 12 | 1288 | 81 |
GenBank mRNA | AK148411.1 | - | - | 1327 | 81 |
AK165677.1 | - | 11 - 12 | 1291 | 81 | |
BC022914.1 | - | 9 - 10 | 2718 | 81 | |
BC027061.1 | - | 11 - 12 | 1313 | 81 |