The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183262.3 | NP_899085.3 | 2 - 3 | 932 | 85 |
GenBank mRNA | AK006778.1 | - | 2 - 3 | 697 | 85 |
BC047277.1 | - | 1 - 2 | 452 | 85 | |
BC138904.1 | - | 2 - 3 | 839 | 85 | |
BC138905.1 | - | 2 - 3 | 836 | 85 |